accessibility menu, dialog, popup

UserName

CAS RN 1073354-32-1

BOOH3CCH3H3CH3CNHNCH3

CAS RN 1073354-32-1

IUPAC Name: N-[(4-methylphenyl)methyl]-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Synonyms: N-[(4-methylphenyl)methyl]-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol): 324.23
Molecular Formula: C19H25BN2O2
InChi Key: AFMJUGQUPACTDK-UHFFFAOYSA-N
SMILES: CC1=CC=C(CNC2=NC=C(C=C2)B2OC(C)(C)C(C)(C)O2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results