CAS RN 105394-83-0
CAS RN 105394-83-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Cyclopropyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one (Decarboxylated Compound), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.