CAS RN 105102-22-5
CAS RN 105102-22-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
9,21-Dichloro-11beta,17-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione (Mometasone), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Mometasone, TRC
CAS: 105102-22-5 Molecular Formula: C22 H28 Cl2 O4 Molecular Weight (g/mol): 427.36 Synonym: Mometasone,Pregna-1,4-diene-3,20-dione, 9,21-dichloro-11,17-dihydroxy-16-methyl-, (11β,16α)- IUPAC Name: (8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-17-(2-chloroacetyl)-11,17-dihydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one SMILES: C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CCl