CAS RN 104953-06-2
CAS RN 104953-06-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
[1-[4-[4-(1,1-Dimethylethyl)-phenyl]but-3-enyl]piperidin-4-yl]diphenylmethanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.