CAS RN 104872-28-8
CAS RN 104872-28-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-Formyl-(D)-leucine (S)-1-[[2S,3S)-3-Hexyl-4-oxo-2-oxetanyl]methyl]dodecyl Ester ((D)-Leucine Orlistat), Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
(R,S,S,S)-Orlistat, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.