CAS RN 104743-23-9
CAS RN 104743-23-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Dimethylaminomethyl-1-phenyl-1-propanol Hydrochloride (alpha-[(Dimethylamino)methyl]-alpha-ethylbenzenemethanol Hydrochloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.