accessibility menu, dialog, popup

UserName

CAS RN 104224-63-7

IUPAC Name: 1-(5-bromo-2-methoxyphenyl)adamantane
Synonyms: 1-(5-bromo-2-methoxyphenyl)adamantane
Molecular Weight (g/mol): 321.26
Molecular Formula: C17H21BrO
InChi Key: QQAMHHZQONQBFZ-UHFFFAOYSA-N
SMILES: COC1=C(C=C(Br)C=C1)C12CC3CC(CC(C3)C1)C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)