accessibility menu, dialog, popup

UserName

ClOHOON

CAS RN 10405-02-4

IUPAC Name: 3-[(2-hydroxy-2,2-diphenylacetyl)oxy]-8λ⁵-azaspiro[bicyclo[3.2.1]octane-8,1'-pyrrolidin]-8-ylium chloride
Synonyms: trospium chloride
Molecular Weight (g/mol): 427.97
Molecular Formula: C25H30ClNO3
InChi Key: RVCSYOQWLPPAOA-UHFFFAOYNA-M
SMILES: [Cl-].OC(C(=O)OC1CC2CCC(C1)[N+]21CCCC1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results