CAS RN 10305-73-4
CAS RN 10305-73-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-1-Aminoindane Hydrochloride, TRC
CAS: 10305-73-4 Molecular Formula: C9 H11 N . Cl H Molecular Weight (g/mol): 169.65 Synonym: 1H-Inden-1-amine, 2,3-dihydro-, hydrochloride (1:1), (1R)-,1-Indanamine, hydrochloride, (R)-(-)- (8CI),1H-Inden-1-amine, 2,3-dihydro-, hydrochloride, (1R)- (9CI),1H-Inden-1-amine, 2,3-dihydro-, hydrochloride, (R)-,(R)-(-)-1-Indanamine hydrochloride,(R)-1-Aminoindan hydrochloride,(R)-1-Aminoindane hydrochloride,(1R)-Indan-1-amine Hydrochloride IUPAC Name: (1R)-2,3-dihydro-1H-inden-1-amine;hydrochloride SMILES: Cl.N[C@@H]1CCc2ccccc12