accessibility menu, dialog, popup

UserName

CAS RN 1019607-55-6

HNN

CAS RN 1019607-55-6

IUPAC Name: 4-(cyclopropylamino)benzonitrile
Synonyms: 4-(cyclopropylamino)benzonitrile
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: SHZXUUVOVQTNCK-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(NC2CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results