accessibility menu, dialog, popup

UserName

CAS RN 1017779-01-9

CH3OOHFFF

CAS RN 1017779-01-9

IUPAC Name: [2-methoxy-6-(trifluoromethyl)phenyl]methanol
Synonyms: [2-methoxy-6-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: UJYWLAALXLVJQH-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1CO)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results