accessibility menu, dialog, popup

UserName

FFFOFFFF

CAS RN 1017777-37-5

IUPAC Name: 2-fluoro-4,6-bis(trifluoromethyl)benzaldehyde
Synonyms: 2-fluoro-4,6-bis(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 260.11
Molecular Formula: C9H3F7O
InChi Key: CXWRFYUFIINRJQ-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(=C1C=O)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results