CAS RN 1016-44-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Alpha,O-Dimethyl Serotonin Hydrochloride, TRC
CAS: 1016-44-0 Molecular Formula: C12 H16 N2 O . Cl H Molecular Weight (g/mol): 240.729 Synonym: 5-Methoxy-α-methyl-1H-indole-3-ethanamine Hydrochloride (1:1),5-Methoxy-α-methyltryptamine Hydrochloride,3-(2-Aminopropyl)-5-methoxyindole Hydrochloride (6CI,7CI,8CI) IUPAC Name: 1-(5-methoxy-1H-indol-3-yl)propan-2-amine;hydrochloride SMILES: Cl.COc1ccc2[nH]cc(CC(C)N)c2c1
2
5-Methoxy-alpha-methyltryptamine Hydrochloride (5-MeO-AMT HCl) 1.0 mg/ml in Methanol (as free base), LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.