CAS RN 101278-21-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Dihydro-3,4-diphenyl-2,5-furandione;, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dihydro-3,4-diphenyl-2,5-furandione;, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dihydro-3,4-diphenyl-2,5-furandione;, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.