accessibility menu, dialog, popup

UserName

ClClBrF

CAS RN 1000572-78-0

IUPAC Name: 1-bromo-4,5-dichloro-2-fluorobenzene
Synonyms: 1-bromo-4,5-dichloro-2-fluorobenzene
Molecular Weight (g/mol): 243.88
Molecular Formula: C6H2BrCl2F
InChi Key: YAKOUNMAKAPKIQ-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=C(Cl)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results