accessibility menu, dialog, popup

UserName

CAS RN 1000546-18-8

CH3FFFOOH

CAS RN 1000546-18-8

IUPAC Name: 2-[2-methyl-3-(trifluoromethyl)phenyl]acetic acid
Synonyms: [2-methyl-3-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 218.18
Molecular Formula: C10H9F3O2
InChi Key: FQVYJURFBWJJPW-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)C=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results