CAS RN 1000342-88-0
IUPAC Name:
3,5-diiodo-6-methylpyridin-2-amine
Synonyms:
3,5-diiodo-6-methylpyridin-2-amine
Molecular Weight (g/mol):
359.94
Molecular Formula:
C6H6I2N2
InChi Key:
BSXUGYBTSPNQDE-UHFFFAOYSA-N
SMILES:
CC1=NC(N)=C(I)C=C1I
1
–
1
of
1
results
1
2-Amino-3,5-diiodo-6-methylpyridine, 95%, Thermo Scientific™
CAS: 1000342-88-0 Molecular Formula: C6H6I2N2 Molecular Weight (g/mol): 359.937 MDL Number: MFCD09880040 InChI Key: BSXUGYBTSPNQDE-UHFFFAOYSA-N Synonym: 2-amino-3,5-diiodo-6-methylpyridine,2-amino-3,5-diiodo-6-methylpridine,3,5-diiodo-6-methyl-pyridin-2-ylamine PubChem CID: 24729486 IUPAC Name: 3,5-diiodo-6-methylpyridin-2-amine SMILES: CC1=NC(=C(C=C1I)I)N