Biuret, 97%
| Quantity | 500 g |
|---|
Chemical Identifiers
| CAS | 108-19-0 |
|---|---|
| Molecular Formula | C2H5N3O2 |
| MDL Number | MFCD00007946 |
| InChI Key | OHJMTUPIZMNBFR-UHFFFAOYSA-N |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| PubChem CID | 7913 |
| ChEBI | CHEBI:18138 |
| IUPAC Name | carbamoylurea |
Specification
| Chemical Name or Material | Biuret |
|---|---|
| CAS | 108-19-0 |
| Melting Point | ∽189°C (decomposition) |
| Density | 1.46 |
| Odor | Odorless |
| Molecular Formula | C2H5N3O2 |
| Linear Formula | H2NCONHCONH2 |
| MDL Number | MFCD00007946 |
| Quantity | 500 g |
| Beilstein | 1703510 |
| Merck Index | 14,1309 |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| Synonym | biuret,allophanamide,imidodicarbonic diamide,carbamylurea,ureidoformamide,dicarbonimidic diamide,allophanic acid amide,allophanimidic acid,dicarbamylamine,isobiuret |
| Solubility Information | Soluble in methanol. Partially soluble in cold water,diethyl ether. |
| InChI Key | OHJMTUPIZMNBFR-UHFFFAOYSA-N |
| SMILES | C(=O)(N)NC(=O)N |
| IUPAC Name | carbamoylurea |
| Molecular Weight (g/mol) | 103.081 |
| PubChem CID | 7913 |
| ChEBI | CHEBI:18138 |
| Formula Weight | 103.08 |
| Percent Purity | 97% |
[Biuret]
[Warning]
[{code=H315, sequence=1, description=Causes skin irritation.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H335, sequence=3, description=May cause respiratory irritation.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P312, sequence=54, description=Call a POISON CENTER/doctor/if you feel unwell.}, {code=P332+P313, sequence=55, description=If skin irritation occurs: Get medical advice/attention.}, {code=P337+P313, sequence=56, description=If eye irritation persists: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: Imidodicarbonic diamide}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
- Biuret
Signal Word
- Warning
Hazard Statement
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
- H335- May cause respiratory irritation.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312- Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313- If skin irritation occurs: Get medical advice/attention.
- P337+P313- If eye irritation persists: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contain: Imidodicarbonic diamide