Learn More
R&L Enterprises™ Zinc Plated Stand Rods
Product Code. 12966806 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1000 mm
Diameter (Metric) Rod:
10 mm
12.5 mm
Unit Size:
Each
Pack of 2
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Rod Unit Size
12966806 1000 mm 12.5 mm Pack of 2
12956806 750 mm 12.5 mm Pack of 2
12368139 600 mm 12.5 mm Pack of 5
11715386 500 mm 10 mm Each
12358139 500 mm 10 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12966806
Estimated Shipment: 12-01-2027
Add to basket
Add to basket
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Zinc Plated Rods are manufactured from mild steel.
  • Threaded
  • Available for a range of diameters and lengths
  • Length of rod does not include the thread
Specifications
Material Zinc Plated Mild Steel
Length (Metric) 1000 mm
Diameter (Metric) Rod 12.5 mm
Product Type Rod