Learn More
Paragon Scientific™ Zeitfuchs Cross-Arm Viscometer for Transparent and Opaque Liquids (UKAS ISO 17025 certified)
For the determination of kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Brand:  Paragon Scientific™ CROSS05UKC
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 16040070
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
  • Supplied with full UKAS ISO 17025 calibration
Specifications
Specifications
60 to 300 cSt
0.3
ASTM D 445, D 446 and ISO 3104, 3105
5
Yes
D 445, D 446
UKAS ISO 17025 calibration
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
8 week lead-time