Learn More
Xylenes, 99.99%, For ACS analysis, MP Biomedicals™
45.80 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Use promo code "23257" to get your promotional price.
Click to view available options
Quantity:
500 mL
Specifications
CAS | 1330-20-7 |
Color | Colorless |
Density | 0.84 g/cm3 at 25°C |
Quantity | 500 mL |
Physical Form | Liquid |
Assay Percent Range | 0.9999 |
Molecular Formula | C8H10 |
Synonym | Xylol |
Formula Weight | 106.17 |
Chemical Name or Material | Xylenes, ACS, For ACS analysis |