Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Double Layered NMR Tubes for Toxic Samples
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Double Layered NMR Tubes provides extra protection when working with toxic samples.
Brand:  Bel-Art™ SP Scienceware™ 66000-0001
Additional Details : Weight : 0.04310kg
 View more versions of this product
Product Code. 15368558
  • 99.05€ / Each
Estimated Shipment: 07-08-2024
Description
  • Inner and outer tubes are designed with an approximately 50 μm gap between them
  • Made of Pyrex™ glass
  • Ideal for variable temperature studies
Specifications
Specifications
Complete Set
2.97 mm
Flat Bottom
5 mm
600 MHz
516-CC-5