Learn More
Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks
Product Code. 15429113 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
250 mL
300 mL
500 mL
1 L
2000 mL
Unit Size:
Each
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15429113 500 mL Pack of 10
15489103 50 mL Pack of 10
15499103 100 mL Pack of 10
11912243 200 mL Pack of 10
15409113 250 mL Pack of 10
15419113 300 mL Pack of 10
15466133 1 L Pack of 10
11813452 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15429113
In Stock
Add to basket
Edge
Add to basket
Constructed from borosilicate glass. Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks are graduated and available in a variety of sizes.
  • Borosilicate glass 3.3
  • Wide Neck
  • Graduated
  • Manufactured to DIN ISO 24450
  • Non-ISO size
Specifications
Certifications/Compliance DIN ISO 24450
Color Transparent
Diameter (English) Outer 105 mm
Diameter (Metric) Base 50 mm
Diameter (Metric) Outer Neck 50 mm
Capacity (Metric) 500 mL
Material Borosilicate Glass (3.3)
Height (Metric) 175 mm
Neck Style Wide
Product Type Erlenmeyer Flask
Shape Conical
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Wide
Sterility Non-sterile
Bottom Shape Flat
Surface Treatment No surface treatment
Quantity 10 PK.