Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Tear-Off Seals
Tear-off seal can be removed completely allowing content to pour from bottle
Brand: DWK Life Sciences 224192-05
109.25 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.30240kg
Description
Tear-off seal can be removed completely allowing content to pour from bottle
- Aluminum
- Unlined
Specifications
Blue | |
1000/Cs. | |
13 mm |
Aluminum | |
Complete Tear-off Cap |