Learn More
DWK Life Sciences™ Wheaton™ Ampule Snapper
Product Code. 10594092 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Plastic
Unit Size:
Pack of 144
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10594092 Plastic Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10594092 Supplier DWK Life Sciences™ Supplier No. 177105
  • ONLINE EXCLUSIVE PRICE
    34.65€
    31.50€ / Pack of 144
    Save 3.15€
    9% Off
In Stock
Add to basket
Edge
Add to basket
Prevent cross-contamination with these snappers for 1 and 2mL ampules
Prevents splintering of ampules
Specifications
Disposable Single-use
Product Type Ampule Snapper
Material Plastic
For Use With (Equipment) 1 and 2mL ampules