missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ PFTE Membrane Filters, TE 37

Chemically stable and inert filters that are suitable for use with aggressive organic solvents, strong acids, and alkalis

Brand:  Cytiva 10411392

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 15601067

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

  • Features polypropylene support material for improved strength and handling
  • Can withstand aggressive solvents, liquids, and gases that damage other membranes
  • Temperature resistant up to 120°C
Specifications

Specifications

Membrane Type: TE 37
1 μm
Hydrophobic
PFTE Membrane Filter
PTFE
5.4 s/100 mL/12.5 cm2
100°C
210 x 297 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers