Learn More
Zwiebel™ Weights Set
Manufactured to the highest standard. Zwiebel™ Weights Set are precise measuring tools.
Brand:  Zwiebel™ PBI56853
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11850982
  • 4277.10€ / Each
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
  • Stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability/In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
E2 Series
Cofrac calibration certificate
Polypropylene
Cylindrical
Calibration Weight
Antimagnetic Stainless Steel
6110 g (Total)
Case includes gripper