Learn More
Abnova™ WDFY2/CEN13q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0176.200uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16043275
  • 2430.00€ / 200µL
Estimated Shipment: 11-06-2024
Specifications
Store at 4°C in the dark.
Tube
115825
200 μL
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe