Learn More
Labconco™ WaterPro™ Filters
Filters, prefilters and cartridges for Labconco WaterPro PS and RO Water Purification Systems.
Brand:  Labconco™ 9007201
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 16297860
  • 184.00€ / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
Activated Carbon Cartridge aids in removal of contaminants during the creation of Type I ultra pure water. For use with Labconco WaterPro PS water Purification Systems.
Specifications
Specifications
Activated Carbon
WaterPro™ PS polishing station Series 90005, 90006 and 90007 model
WaterPro Filters
1.8 kg