Learn More
Labconco™ WaterPro™ Filters
Filters, prefilters and cartridges for Labconco WaterPro PS and RO Water Purification Systems.
Brand: Labconco™ 9007201
Additional Details : Weight : 2.00000kg
Description
Activated Carbon Cartridge aids in removal of contaminants during the creation of Type I ultra pure water. For use with Labconco WaterPro PS water Purification Systems.Specifications
Activated Carbon | |
WaterPro™ PS polishing station Series 90005, 90006 and 90007 model | |
WaterPro Filters | |
1.8 kg |