Learn More
Labconco™ WaterPro™ Filters
Filters, prefilters and cartridges for Labconco WaterPro PS and RO Water Purification Systems.
Brand: Labconco™ 9053300
Additional Details : Weight : 2.00000kg
Description
Organic Adsorption Cartridge aids in removal of organic contaminants during the creation of Type I ultra pure water. For use with WaterPro PS models.Specifications
Organic adsorption cartridge | |
WaterPro™ PS/HPLC polishing station (Labconco Cat. Nos. 90006-00 to -03) and HPLC/UF Hybrid (Labconco Cat. Nos. 90007-04, -05) | |
WaterPro Filters | |
1.8 kg |