Learn More
Labconco™ WaterPro™ Filters
Filters, prefilters and cartridges for Labconco WaterPro PS and RO Water Purification Systems.
Brand:  Labconco™ 9053300
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 16217870
  • 314.00€ / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
Organic Adsorption Cartridge aids in removal of organic contaminants during the creation of Type I ultra pure water. For use with WaterPro PS models.
Specifications
Specifications
Organic adsorption cartridge
WaterPro™ PS/HPLC polishing station (Labconco Cat. Nos. 90006-00 to -03) and HPLC/UF Hybrid (Labconco Cat. Nos. 90007-04, -05)
WaterPro Filters
1.8 kg