Learn More
Labconco™ WaterPro™ Filters
Filters, prefilters and cartridges for Labconco WaterPro PS and RO Water Purification Systems.
Brand:  Labconco™ 9007301
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 16207870
  • 274.00€ / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
Deionization Cartridge is polishing grade mixed bed to deliver 18.2 megohm-cm quality water with TOC levels below 5 ppb.
Specifications
Specifications
Deionization cartridge
WaterPro™ PS polishing station Series 90005, 90006 and 90007 model
WaterPro Filters
1.8 kg