Learn More
Viral Production Medium, prototype
Viral Production Medium, prototype, requires supplementation with 4 mM GlutaMAX Supplement prior to use.
Brand: Invitrogen A49842DK
Additional Details : Weight : 1.11000kg
Description
Gibco Viral Production Medium, prototype, is a chemically defined, serum-free, protein-free medium specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells) for adeno-associated virus (AAV) production as part of the Gibco AAV-MAX Helper-Free AAV production system.
Specifications
Store at 2–8°C, protected from light. | |
Mammalian Cells | |
Mammalian | |
GlutaMAX | |
AAV Production | |
RUO |
HEK293 | |
Suspension Cell Culture | |
Liquid | |
Bottle | |
AAV-MAX |
For Research Use Only. Not for use in diagnostic procedures. This product is a prototype and performance characteristics of this product have not been established.