Learn More
Beskrivning
- For use with S-950 Premium Head Suspension
- Wrapped around headband to create more comfortable fit and to absorb sweat
- 3M Catalog# S-953 , 3M Catalog# S-954
Specifications
Specifications
| Color | Gray |
| Disposable | Reusable |
| Product Type | Head Suspension |
| Size | Small |
| For Use With (Application) | Cleaning, Electrical, Facility Maintenance, Welding |
| For Use With (Equipment) | Premium Head Suspension |
| Model | S-954 |