Learn More
Cole-Parmer™ VapLock™ Activated Media Exhaust Filters
VapLock™ Activated Media Exhaust Filters reduce harmful chemical emissions and odors from solvent containers
Brand: Cole-Parmer™ 12019-80
Additional Details : Weight : 0.01000kg
Description
Use with indicators to set a replacement scheduleServices
None
Specifications
Solvent Safety | |
Activated Carbon with Breakthrough Indicator | |
1/4 in. NPT(F) |
Exhaust Filter with Indicator, 75 g activated carbon, 1/4 in. NPT(F); 1/ea | |
VapLock Solvent Containment Systems |