Learn More
Cole-Parmer™ VapLock™ Activated Media Exhaust Filters
VapLock™ Activated Media Exhaust Filters reduce harmful chemical emissions and odors from solvent containers
Brand:  Cole-Parmer™ 12019-80
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16682033
  • 125.00€ / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
Use with indicators to set a replacement schedule
Services
None
Specifications
Specifications
Solvent Safety
Activated Carbon with Breakthrough Indicator
1/4 in. NPT(F)
Exhaust Filter with Indicator, 75 g activated carbon, 1/4 in. NPT(F); 1/ea
VapLock Solvent Containment Systems