Learn More
Thermo Scientific™ Vacutherm™ Oven Accessories and Options
Choose from a range of accessories and options for Vacutherm™ ovens to meet application-specific requirements.
Brand: Thermo Scientific™ 51900079
1160.61 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
Choose from a range of Thermo Scientific™ Vacutherm™ Oven Accessories and Options to meet application-specific requirements.
Spécification
Rated temperature of 400°C (door without inspection window) | |
VT 6060 P | |
400°C | |
Door |