Learn More
Integra Biosciences™ Vacusafe™ Comfort Aspiration system
Product Code. 10590273
Change view
Click to view available options
Type:
Vacusafe Comfort Aspiration System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10590273 Vacusafe Comfort Aspiration System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10590273 Supplier Integra Biosciences™ Supplier No. 158310
  • ONLINE EXCLUSIVE PRICE
    2919.02€
    2342.93€ / Pack of 1
    Save 576.09€
    19% Off
View all promotions
Estimated Shipment: 17-11-2026
Add to basket
Edge
Add to basket
Collect and contain biological liquid waste. Integra Biosciences™ Vacusafe™ Comfort Aspiration system is a compact laboratory aspiration system.
  • Compact
  • Level detection sensor
  • Control knob for infinitely vacuum setting (range: -300mbar to 600mbar)
  • Silent operation.
  • All parts exposed to critical liquids are fully autoclavable and can be sterilized for 20 minutes at 121°C,
  • Liquids are collected in the autoclavable waste bottle.
  • Hydrophobic filter reduces the risk of accidently aspirating liquid into the pump and pump housing.
  • Available with either a 4L polypropylene bottle or a 3L glass bottle. 
Specifications
Includes 4 Liter Bottle, Level detection and lid with quick tubing couplings (PVDF)
Quantity 1 Each
Type Vacusafe Comfort Aspiration System