Learn More
Sperian™ V-Maxx Acetate Safety Goggle
8.23 EUR valid until 2024-03-30
Use promo code "21636" to get your promotional price.
Use promo code "21636" to get your promotional price.
Click to view available options
Lens Tint:
Clear
Specifications
Certifications/Compliance | ISO 9001 / 2000 |
Lens Coating | Anti-fog |
Lens Tint | Clear |
Product Type | Safety goggles |
For Use With (Application) | Painting, General Use and Protection Against Fogging |
Headband Material | Neoprene |
No. per Pack | 1 Ea. |
Ventilation Details | Indirect |
Material | Acetate |