Learn More
Fisherbrand™ pH Indicator Paper Strips
Product Code. 15930872 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
1 to 11
1 to 14
Unit Size:
4 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
15930872 1 to 14 4 units
15920872 1 to 11 4 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15930872
  • ONLINE EXCLUSIVE PRICE
    30.96€
    20.68€ / Pack of 4
    Save 10.28€
    33% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Provides optimal quality and reliable readings. Fisherbrand™ pH Indicator Paper Strips are supplied in plastic reels that ensure long-term stability and protection against many external influences.
  • Ready to use
  • Manufactured from high quality filter paper
  • Colors of the color scales are specially mixed to perfectly match the reaction color of the indicator papers
Specifications
pH Range 1 to 14
Format Roll
Quantity 4 reels
Type pH Paper
Length (Metric) 5 m
Width (Metric) 7 mm