Learn More
Fisherbrand™ Universal pH Indicator Paper
Product Code. 11763554 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
0 to 10
1 to 14
5 to 9
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
11763554 1 to 14 Each
11703564 5 to 9 Each
11783554 0 to 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11763554
  • ONLINE EXCLUSIVE PRICE
    5.75€
    4.74€ / Pack of 1
    Save 1.01€
    17% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
  • Filter paper soaked with one or several colored indicators of pH
  • Dip indicator paper into the liquid to be tested for a few seconds and remove it from the test solution and compare the resultant colors with the color scale printed on the cover
  • Provided on a dispenser roll of 5m
  • Two pH ranges available: 1 to 14 and 1 to 11
Specifications
pH Range 1 to 14
For Use With (Application) Measuring pH
Quantity 5 m Dispenser Roll
Test Method Indicator strips