Learn More
Fisherbrand™ X3 Universal Indicator pH Paper Roll Refill, pH 1-14
Product Code. 10151622 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
1 to 14
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10151622 1 to 14 Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10151622
  • ONLINE EXCLUSIVE PRICE
    15.85€
    13.67€ / Pack of 3
    Save 2.18€
    13% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Quickly and easily measure solution pH value at the point of interest with this paper impregnated with universal indicator solution. Fisherbrand™ Universal Indicator pH Paper Roll exhibits smooth color changes over a pH value range from 1 to 14.
  • Ready to use.
  • No calibration, set-up, or maintenance.
  • Perfect for a variety of environments.
Order Info
This is a refill for Fisherbrand Universal pH Indicator Paper, Range 1-14 (Code 11763554).
Specifications
pH Range 1 to 14
Quantity 1 Roll
Type Universal pH Indicator Paper - Refill, grade 911