Learn More
Abeba™ UNI6 6771 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand:  Abeba™ 6771/36
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15662198
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20347:2012 O2 FO SRC
  • Smooth leather
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Specifications
CE EN ISO 20347:2012 O2 FO SRC
Leather
Slip-resistant
6771
Black
36
Polyurethane