Learn More
Abeba™ UNI6 6701 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand:  Abeba™ 6701/40
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15601998
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRC
  • Velours with breathable textile inlays
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • Antistatic
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Reflective material
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRC
Microfiber
40
Polyurethane
Black
Velour
Slip-resistant
6701