Learn More
Abeba™ UNI6 1782 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
Brand: Abeba™ 1782/41
Additional Details : Weight : 1.12000kg
Description
- CE EN ISO 20345:2011 S2 SRC
- A-micro
- Gray/Blue
- Steel toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350122)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- **Size 35 with replaceable acc Wave insole (art. 3576)
Specifications
CE EN ISO 20345:2011 S2 SRC | |
A-micro | |
Slip-resistant | |
1782 |
Gray, Blue | |
41 | |
Polyurethane | |
Steel |