Learn More
Azpack™ Sarogold™ PRO Cling Wrap Film
Product Code. 11958462
Change view
Click to view available options
Width (Metric):
300mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
11958462 300mm Each
12872233 300mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11958462 Supplier Azpack™ Supplier No. SARAN
  • ONLINE EXCLUSIVE PRICE
    112.00€
    108.78€ / Pack of 1
    Save 3.22€
    2% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Provides a total barrier to both moisture and gases preventing contamination of the silicone rubber seals of the gel dryer. Azpack™ Sarogold™ PRO Cling Wrap Film extracts vapours downwards through the gel to prevent uneven drying.
  • For use with gel drying applications
  • Provides an impervious barrier
  • Readily removes from dried gel surface without lifting the gel with it.
  • Clear, ultra violet transparent and may be used to cover and store pre-cast gels for a considerable period at +4°C, or to cover agarose gels preventing drying during observation and photography on a transilluminator.
  • Supplied in a 300m roll in stiff cardboard, cutter boxes.
Specifications
Color Clear
Product Type Cling Film
Width (Metric) 300mm
Material Plastic