Learn More
BRAND™ Ultra Low Retention Filter Tips
Significantly reduce sample loss and provide substantially higher reproducibility when working with critical media.
Brand: BRAND™ 732804
9.01 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Additional Details : Weight : 0.36000kg
Description
- Cleanroom quality for contamination-free analyses
- Perfect fit for the Transferpette™ micropipette and many other pipettes
- Polypropylene construction with polyethylene filter
- TipBox packaging
- Non-sterile
- Autoclavable at 121°C
- DIN EN 285 and CE-marked according to IVD-Directive 98/79 EC
- DNA-, RNase-, endotoxin-free
Specifications
Ultra Low Retention | |
Filtered | |
Natural | |
CE-IVD, DE-M | |
TipBox | |
Gray | |
Eppendorf™ Research™, Eppendorf™ Reference™ | |
ULR | |
2, 10 μL |
0.5 to 10 μL | |
Non-sterile | |
Autoclavable | |
Hinged Rack | |
480/PK | |
Single-use | |
Yes | |
Polypropylene, Polyethylene (Filter) |