Learn More
Brady™ Ultra Fine Pen
Product Code. 16449015 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16449015 Black Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16449015 Supplier Brady™ Supplier No. 335092
  • ONLINE EXCLUSIVE PRICE
    29.45€
    24.85€ / Pack of 5
    Save 4.60€
    15% Off
In Stock
Add to basket
Add to basket
Labeling pen uses a quick drying, permanent, water-resistant ink.
  • Our label markers use permanent, quick-drying, water-resistant ink
Specifications
Alternate Part Number BFS-10
Product Type Marker
Color Black
For Use With (Application) Labeling, General identification