Learn More
Type B Microsyringe
Brand:  Hospidex MSE05R
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11759823
  • 141.00€ / Each
Estimated Shipment: 18-06-2024
Specifications
0.715 mm
1/Pk.
51 mm
Microsyringe
5 μL
Fixed