missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-PHOENIX Touchless-90 Warning Yellow

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3012000233715

Additional Details : Weight : 425.95920kg

 View more versions of this product

Product Code. 15922756

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Space saving folding doors: less required space in front of the cabinet while doors are open
  • Effortless use: the sensor detects a person within approx. 2 seconds
  • User-friendly and safe: after detection, the folding doors open automatically within approx. 5 seconds. The sensor recognises the person while operating and hold the folding doors open
  • Always closed: if the person leaves the sensor-controlled detection area, the folding doors close automatically after approx. 30 seconds
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing  locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

1742 mm
Safety storage cabinet
1050 mm
Powder-coated plain
1196 mm
473.50 kg
Yellow
616 mm
Folding, Automatic opening
3
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
524 mm
Steel
616 x 1196 x 1968 mm
524 x 1050 x 1742 mm
Standard
1968 mm
S90.196.120.FDAO
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers